SY3465084895 Synthonix Corporation

methyl 5-chloroimidazo[1,5-a]pyridine-6-carboxylate

  • MDL number MFCD23711382
  • Cas Number : 1168090-92-3
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/16929.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/16929.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart