SY3465084927 Synthonix Corporation

1-methyl-4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]piperidine

  • MDL number MFCD21337767
  • Cas Number : 1323919-64-7
  • Molecular Weight : 291.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/18647.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/18647.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart