SY3465085099 Synthonix Corporation

{2-methyl-2H-pyrazolo[3,4-b]pyridin-5-yl}boronic acid

  • MDL number MFCD20926164
  • Cas Number : 2304634-03-3
  • Molecular Weight : 176.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/27903.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/27903.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart