SY3465085128 Synthonix Corporation

1h-pyrazolo[4,3-b]pyridine-6-carbonitrile

  • MDL number MFCD20923317
  • Cas Number : 1352395-13-1
  • Molecular Weight : 144.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/18884.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/18884.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart