SY3465085187 Synthonix Corporation

sodium (2Z)-1,4-diethoxy-1,4-dioxobut-2-en-2-olate

  • MDL number MFCD00035571
  • Cas Number : 40876-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/23572.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/23572.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart