SY3514241716 Synthonix Corporation

tert-butyl (3R,4S)-4-hydroxy-3-methyl-piperidine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 955028-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/30027.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/30027.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart