SY3514241723 Synthonix Corporation

methyl 3-[(3-aminocyclobutanecarbonyl)amino]cyclobutanecarboxylate hydrochloride

  • MDL number MFCD30146437
  • Cas Number : 1860028-26-7
  • Molecular Weight : 299.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/30136.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/30136.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart