SY3514241725 Synthonix Corporation

tert-butyl N-{[(2R,4S)-4-fluoropyrrolidin-2-yl]methyl}carbamate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1818843-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/30163.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/30163.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart