SY3514241733 Synthonix Corporation

5-ethenyl-2-fluoro-pyridine

  • MDL number MFCD18384201
  • Cas Number : 1133879-66-9
  • Molecular Weight : 123.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/16703.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/16703.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart