SY3514241765 Synthonix Corporation

[3-(trifluoromethyl)pyridin-4-yl]boronic acid

  • MDL number No MDL found
  • Cas Number : 1204334-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17431.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/17431.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart