SY3514241777 Synthonix Corporation

[1,2,4]triazolo[1,5-a]pyridine-7-carboxylic acid

  • MDL number MFCD17015981
  • Cas Number : 1234616-29-5
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/17857.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/17857.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart