SY3514241780 Synthonix Corporation

2-chloro-3H,4H,7H-pyrrolo[2,3-d]pyrimidin-4-one

  • MDL number MFCD23702052
  • Cas Number : 1245811-22-6
  • Molecular Weight : 169.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/17963.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/17963.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart