SY3514241947 Synthonix Corporation

tert-butyl N-[(3S,4S)-3-hydroxypiperidin-4-yl]carbamate

  • MDL number MFCD22384728
  • Cas Number : 724788-22-1
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/25073.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/25073.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart