SY3514241975 Synthonix Corporation

(2R)-2-methyl-1-[(1S)-1-phenylethyl]piperidin-4-one

  • MDL number MFCD12031215
  • Cas Number : 89467-37-8
  • Molecular Weight : 217.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/26270.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/26270.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart