SY3514241991 Synthonix Corporation

(1R)-1-(pyrimidin-5-yl)ethan-1-amine hydrochloride

  • MDL number MFCD28166335

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/27095.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/27095.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart