SY3514242041 Synthonix Corporation

Thieno[2,3-b]pyridine-5-carboxylic acid

  • MDL number MFCD19690393
  • Cas Number : 117390-38-2
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29526.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29526.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart