SY3514242053 Synthonix Corporation

5-Bromo-6-chloro-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD12963482
  • Cas Number : 1190321-59-5
  • Molecular Weight : 231.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29541.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29541.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart