SY3514242054 Synthonix Corporation

4-Bromo-5-methoxy-1H-indazole

  • MDL number MFCD17676852
  • Cas Number : 1192004-62-8
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29542.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29542.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart