SY3514242111 Synthonix Corporation

Benzatropine mesilate

  • MDL number MFCD00074784
  • Cas Number : 132-17-2
  • Molecular Weight : 403.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29611.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29611.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart