SY3514242138 Synthonix Corporation

ethyl 4-amino-3,5-difluorobenzoate

  • MDL number MFCD28678682
  • Cas Number : 1415920-00-1
  • Molecular Weight : 201.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29649.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29649.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart