SY3514242165 Synthonix Corporation

methyl 6-bromo-3-methyl-1H-indole-4-carboxylate

  • MDL number MFCD27952312
  • Cas Number : 1583272-35-8
  • Molecular Weight : 268.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29687.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29687.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart