SY3514242227 Synthonix Corporation

O1-tert-butyl O2-methyl (2S,4S)-4-methoxypyrrolidine-1,2-dicarboxylate

  • MDL number MFCD16621201
  • Cas Number : 215918-38-0
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29767.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29767.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart