SY3514242249 Synthonix Corporation

(2R,3R)-1-[(tert-butoxy)carbonyl]-3-hydroxypyrrolidine-2-carboxylic acid

  • MDL number MFCD26406357
  • Cas Number : 335280-19-8
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29807.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29807.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart