SY3514242259 Synthonix Corporation

tert-butyl (3R,4S)-rel-3-hydroxy-4-methylpiperidine-1-carboxylate

  • MDL number MFCD18791214
  • Cas Number : 374794-75-9
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29825.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29825.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart