SY3514242322 Synthonix Corporation

5-bromo-1-{[2-fluoro-6-(trifluoromethyl)phenyl]methyl}-6-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD12546646
  • Cas Number : 830346-48-0
  • Molecular Weight : 381.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29944.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29944.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart