SY3514242391 Synthonix Corporation

N-Methyl-N-[(3S)-pyrrolidin-3-yl]methanesulfonamide hydrochloride

  • MDL number MFCD29037480
  • Cas Number : 1810074-78-2
  • Molecular Weight : 214.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/30040.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/30040.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart