SY3514242395 Synthonix Corporation

methyl 6-bromo-1-methyl-2-oxo-indoline-4-carboxylate

  • MDL number MFCD29060180
  • Cas Number : 1818847-70-9
  • Molecular Weight : 284.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/30044.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/30044.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart