SY3514242489 Synthonix Corporation

tert-butyl 3-(2-methoxy-2-oxoethyl)-4-methylpiperidine-1-carboxylate

  • MDL number MFCD29060205
  • Cas Number : 1860028-35-8
  • Molecular Weight : 271.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/30138.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/30138.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart