SY3638138162 Synthonix Corporation

2-Chloro-4-fluoro-benzylhydrazine dihydrochloride

  • MDL number MFCD30489782
  • Cas Number : 2044707-02-8
  • Molecular Weight : 247.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/31594.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/31594.html
pictograms GHS07
hstatements H313, H315, H320
Availability details for
Quantity
  • Availability
  • Add to Cart