SY3638138170 Synthonix Corporation

2-Chloromethyl-5-fluoropyrimidine hydrochloride

  • MDL number MFCD22570242
  • Cas Number : 2052356-33-7
  • Molecular Weight : 183.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/31600.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/31600.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart