SY3638138184 Synthonix Corporation

1-Methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

  • MDL number MFCD04114000
  • Cas Number : 1020174-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/16052.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/16052.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart