SY3786986282 Synthonix Corporation

1,3-Dihydropyrrolo[2,3-c]pyridin-2-one hydrochloride

  • MDL number MFCD08234948
  • Cas Number : 295327-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0383.html

Safety Information

pcodes P280 ,
MSDS https://synthonix.com/0383.html
hstatements H301 , H319 ,
Availability details for
Quantity
  • Availability
  • Add to Cart