SY3786986302 Synthonix Corporation

2,5-Diazabicyclo[2.2.2]octane-2-carboxylic acid tert-butylester hemioxalate

  • MDL number MFCD29037485
  • Cas Number : 1810070-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/6591.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/6591.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart