SY3786986397 Synthonix Corporation

endo-3-(BOC-amino)-8-azabicyclo[3.2.1]octane

  • MDL number MFCD16660224
  • Cas Number : 132234-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6248.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6248.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart