SY3786986401 Synthonix Corporation

tert-Butyl 7-oxa-3-aza-bicyclo[4.1.0]heptane-3-carboxylate

  • MDL number MFCD12407050
  • Cas Number : 161157-50-2
  • Molecular Weight : 199.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97.00
Availability details for
Quantity
  • Availability
  • Add to Cart