SY3H11AB1F56 Synthonix Corporation

5-Aminopentanenitrile hydrochoride

  • MDL number MFCD23152701
  • Cas Number : 1638108-03-8
  • Molecular Weight : 134.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6983.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6983.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart