SY3H11AB1FCB Synthonix Corporation

2-(1,3-Dioxolan-2-yl)phenylmagnesium bromide, 0.5 M in THF

  • MDL number MFCD02260186
  • Cas Number : 137964-68-2
  • Molecular Weight : 253.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/31798.html

Safety Information

signalword Danger
pcodes P222, P223, P231, P232, P261, P280
MSDS https://synthonix.com/31798.html
pictograms GHS02, GHS05
hstatements H225, H302, H314, H319, H335, H351
Availability details for
Quantity
  • Availability
  • Add to Cart