SY3H30324D71 Synthonix Corporation

5-(Cyclopropylmethoxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzodioxole

  • MDL number MFCD11706989
  • Cas Number : 1185836-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/4959.html

Safety Information

MSDS https://synthonix.com/4959.html
Availability details for
Quantity
  • Availability
  • Add to Cart