SY3H30324D85 Synthonix Corporation

tert-butyl-(3-Iodoprop-2-ynoxy)dimethylsilane

  • Cas Number : 159009-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/1889.html

Safety Information

MSDS https://synthonix.com/1889.html
Availability details for
Quantity
  • Availability
  • Add to Cart