SY3H30324D8D Synthonix Corporation

1-Butoxy-3-methoxybenzene

  • MDL number MFCD31699842
  • Cas Number : 51241-32-8
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/4970.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/4970.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart