SY3H30324D93 Synthonix Corporation

2-Bromo-1-(cyclopropylmethoxy)-4-methylbenzene

  • Cas Number : 1154340-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/5008.html

Safety Information

MSDS https://synthonix.com/5008.html
Availability details for
Quantity
  • Availability
  • Add to Cart