SY3H30324DA1 Synthonix Corporation

Ethyl 2-(2-but-3-ynyl-1,3-dioxolan-2-yl)acetate

  • MDL number No MDL found
  • Cas Number : 91798-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6253.html

Safety Information

MSDS https://synthonix.com/6253.html
Availability details for
Quantity
  • Availability
  • Add to Cart