SY3H30324DA2 Synthonix Corporation

Ethyl 3-oxohept-6-ynoate

  • MDL number MFCD24688431
  • Cas Number : 35116-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/6254.html

Safety Information

MSDS https://synthonix.com/6254.html
Availability details for
Quantity
  • Availability
  • Add to Cart