SY3H30324DA6 Synthonix Corporation

1-Fluoro-3-((3-methoxyphenoxy)methyl)benzene

  • MDL number MFCD31699845
  • Cas Number : 850461-60-8
  • Molecular Weight : 232.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/4963.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/4963.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart