SY3H30324DA8 Synthonix Corporation

trans-3-Hydroxy-3-methylcyclobutane-1-methamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 2231664-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/31870.html

Safety Information

MSDS https://synthonix.com/31870.html
Availability details for
Quantity
  • Availability
  • Add to Cart