SY3H30324DAE Synthonix Corporation

(Iodoethynyl)triisopropylsilane

  • MDL number MFCD28015634
  • Cas Number : 160481-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/2773.html

Safety Information

MSDS https://synthonix.com/2773.html
Availability details for
Quantity
  • Availability
  • Add to Cart