SY3H30324DB3 Synthonix Corporation

9-Methyl-9H-fluoren-9-ol

  • MDL number No MDL found
  • Cas Number : 6311-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/4999.html

Safety Information

MSDS https://synthonix.com/4999.html
Availability details for
Quantity
  • Availability
  • Add to Cart