SY3H304528A9 Synthonix Corporation

7-Bromo-4,4-dimethyl-1H-benzo[d][1,3]oxazin-2(4H)-one

  • Cas Number : 1245643-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32013.html

Safety Information

MSDS https://synthonix.com/32013.html
Availability details for
Quantity
  • Availability
  • Add to Cart