SY3H304528AA Synthonix Corporation

6-Bromoquinolin-8-ol

  • Cas Number : 139399-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32018.html

Safety Information

MSDS https://synthonix.com/32018.html
Availability details for
Quantity
  • Availability
  • Add to Cart