SY3H304528BD Synthonix Corporation

4-Bromo-2-nitropyridine

  • Cas Number : 909712-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32113.html

Safety Information

MSDS https://synthonix.com/32113.html
Availability details for
Quantity
  • Availability
  • Add to Cart